Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDCD2 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306434G
This item is not returnable.
View return policy
Description
NUDCD2 Polyclonal antibody specifically detects NUDCD2 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
NUDCD2 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
DyLight 488 | |
DKFZp686E10109, NudC domain containing 2, nudC domain-containing protein 2, NudC-like protein 2 | |
Produced in rabbits immunized with E. coli-derived Human NUDCD2 fragment. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
134492 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction