Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDCD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546769
Description
Peptide sequence: YAADPWVQDQ MQRKLTLERF QKENPGFDFS GAEISGNYTK GGPDFSNLEK.
NUDCD2 may regulate the LIS1/dynein pathway by stabilizing LIS1 with Hsp90 chaperone.
Specifications
| NUDCD2 | |
| Polyclonal | |
| Unconjugated | |
| Nudcd2 | |
| D11Ertd603e; NudC domain containing 2; nudC domain-containing protein 2; Nudcd2; NudCL2; NudC-like protein 2; RGD1307203 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 134492 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WVJ2 | |
| Nudcd2 | |
| Synthetic peptide directed towards the C-terminal of human NUDCD2 (aa 108-157). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction