Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDCD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553936
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553936 Supplier Invitrogen™ Supplier No. PA553936
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRKEEEEAKT VSAAAAEKEP VPVPVQEIEI DSTTELDGHQ EVEKVQPPGP VKEMAHGSQE AEAPGAVAGA AE Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 63%.

NUDCD3 gene ontology annotations related to this gene include cilium assembly; developmental process; protein folding; protein localization to pericentriolar material.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDCD3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUDCD3
Gene Accession No. Q8IVD9
Gene Alias AI427847; BC024322; KIAA1068; KIAA1068 protein; mKIAA1068; nudC domain containing 3; nudC domain-containing protein 3; nudcd3; NudCL; NudC-like protein; Unknown (protein for MGC:137347); zgc:77067
Gene Symbols NUDCD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUDCD3. Recombinant protein control fragment (Product #RP-92610).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23386
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.