Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDCD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544297
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544297 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544297 Supplier Invitrogen™ Supplier No. PA544297
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KINKERSMAT VDEEEQAVLD RLTFDYHQKL QGKPQSHELK VHEMLKKGWD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

NUDCD3 gene ontology annotations related to this gene include cilium assembly; developmental process; protein folding; protein localization to pericentriolar material.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDCD3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NUDCD3
Gene Accession No. Q8IVD9
Gene Alias AI427847; BC024322; KIAA1068; KIAA1068 protein; mKIAA1068; nudC domain containing 3; nudC domain-containing protein 3; nudcd3; NudCL; NudC-like protein; Unknown (protein for MGC:137347); zgc:77067
Gene Symbols NUDCD3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human NUDCD3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23386
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.