Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDCD3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501727437 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7437 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7437 Supplier Proteintech Group Inc Supplier No. 117641AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUDCD3 gene ontology annotations related to this gene include cilium assembly; developmental process; protein folding; protein localization to pericentriolar material.

Specifications

Antigen NUDCD3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NUDCD3
Gene Accession No. Q8IVD9, Q8R1N4
Gene Alias KIAA1068, NudC domain containing 3, NUDCD3, NudCL
Gene Symbols Nudcd3
Host Species Rabbit
Immunogen NUDCD3 Fusion Protein Ag2362
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209586, 23386
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.