Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nudel Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596422
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596422 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596422 Supplier Invitrogen™ Supplier No. PA596422
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDGEDIPDFS SLKEETAYWK ELSLKYKQSF QEARDELVEF QEGSRELEAE LEAQLVQAEQ RNRDLQADNQ RLKYEVEALK EKLEHQYAQS YKQVSVLEDD LSQTRAIKEQ LHKYVRELEQ ANDDLERAKR ATIVSLEDFE QRLNQAIERN AFLESELDEK ESLLVSVQRL KDEARDLRQE LAVRERQQEV TRKSAPSSPT LDCEKMDSAV QASLSLPATP VGKGTENTFP SPKAIPNGFG TSPLTPSARI SALNIVGDLL RKVGALESKL AACRNFAKDQ ASRKSYISGN VNCGVLNGNG TKFSRSGHTS FFDKGAVNGF DPAPPPPGLG SSRPSSAPGM LPLSV; Positive Samples: U-87MG, Mouse brain; Cellular Location: Chromosome, Cytoplasm, centromere, centrosome, cytoskeleton, kinetochore, microtubule organizing center, spindle.

This gene encodes a thiol-activated peptidase that is phosphorylated in M phase of the cell cycle. Phosphorylation regulates the cell cycle-dependent distribution of this protein, with a fraction of the protein bound strongly to centrosomes in interphase and localized to mitotic spindles in early M phase. Overall, this protein plays a role in nervous system development. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nudel
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ndel1
Gene Accession No. Q9ERR1, Q9GZM8
Gene Alias 2600006O07Rik; ATP-sensitive inward rectifier potassium channel 15; DKFZp451M0318; endooligopeptidase A; EOPA; Inward rectifier K(+) channel Kir4.2; Kcnj15; Kir4.2; LIS1-interacting protein NUDEL; LIS1-interacting protein NUDEL; endooligopeptidase A; MITAP1; Mitosin-associated protein 1; mitosin-associated protein MITAP1; mNudel; mNudE-L; NDE1L1; NDE2; Ndel1; nuclear disribution gene E like; nuclear distribution gene E-like homolog 1; nuclear distribution gene E-like homolog 1 (A. nidulans); nuclear distribution protein nudE-like 1; nudE neurodevelopment protein 1 like 1; nudE neurodevelopment protein 1-like 1; nudE nuclear distribution E homolog-like 1; nudE nuclear distribution gene E homolog like 1; nudE nuclear distribution gene E homolog-like 1; Nude2; NUDEL; NUDE-like protein; Potassium channel, inwardly rectifying subfamily J member 15; potassium channel, inwardly rectifying subfamily J, member 15; potassium inwardly-rectifying channel, subfamily J, member 15; Protein mNudE-like; Protein Nudel; unnamed protein product
Gene Symbols Ndel1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-345 of human NUDEL/NUDEL/NDEL1 (NP_1104351).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81565, 83431
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.