Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDT10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA551322
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA551322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA551322 Supplier Invitrogen™ Supplier No. PA551322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total NUDT10 protein.

Cleaves a beta-phosphate from the diphosphate groups in PP-InsP5 (diphosphoinositol pentakisphosphate), suggesting that it may play a role in signal transduction. Also able to catalyze the hydrolysis of dinucleoside oligophosphates, with Ap6A and Ap5A being the preferred substrates. The major reaction products are ADP and p4a from Ap6A and ADP and ATP from Ap5A. Also able to hydrolyze 5-phosphoribose 1-diphosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene NUDT10
Gene Accession No. Q8NFP7
Gene Alias AI853080; APS2; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-alpha; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-beta; Diadenosine hexaphosphate hydrolase (AMP-forming); diphosphoinositol polyphosphate phosphohydrolase 3-alpha; Diphosphoinositol polyphosphate phosphohydrolase 3-beta; DIPP3 alpha; Dipp3a; DIPP-3-alpha; DIPP3-alpha; Dipp3b; DIPP-3-beta; hAps2; hDIPP3alpha; MNCb-1696; Nucleoside diphosphate-linked moiety X motif 10; Nucleoside diphosphate-linked moiety X motif 11; nudix (nucleoside diphosphate linked moiety X)-type motif 10; nudix hydrolase 10; nudix motif 10; Nudix motif 11; Nudt10; Nudt11; RGD1564763
Gene Symbols NUDT10
Host Species Rabbit
Immunogen Full length fusion protein of human NUDT10.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170685
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.