Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT11 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306300AF750
Description
NUDT11 Polyclonal specifically detects NUDT11 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
NUDT11 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
APS1DIPP3B, ASP1, Diadenosine 5'-5'''-P1, diphosphoinositol polyphosphate phosphohydrolase 3, beta, diphosphoinositol polyphosphate phosphohydrolase 3-beta, DIPP3b, DIPP3beta, DIPP-3-beta, DIPP3-beta, EC 3.6.1.-, EC 3.6.1.52, FLJ10628, hDIPP3betahAps1, Nucleoside diphosphate-linked moiety X motif 11, nudix (nucleoside diphosphate linked moiety X)-type motif 11, Nudix motif 11, P6-hexaphosphate hydrolase 3-beta | |
Produced in rabbits immunized with E. coli-derived Human NUDT11 fragment. | |
0.1 mL | |
Signal Transduction | |
55190 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction