Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDT10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563467
Description
Immunogen sequence: LQCHKPVHAE YLEKLKLGGS PTNGNS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Cleaves a beta-phosphate from the diphosphate groups in PP-InsP5 (diphosphoinositol pentakisphosphate), suggesting that it may play a role in signal transduction. Also able to catalyze the hydrolysis of dinucleoside oligophosphates, with Ap6A and Ap5A being the preferred substrates. The major reaction products are ADP and p4a from Ap6A and ADP and ATP from Ap5A. Also able to hydrolyze 5-phosphoribose 1-diphosphate.
Specifications
NUDT10 | |
Polyclonal | |
Unconjugated | |
NUDT10 | |
AI853080; APS2; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-alpha; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-beta; Diadenosine hexaphosphate hydrolase (AMP-forming); diphosphoinositol polyphosphate phosphohydrolase 3-alpha; Diphosphoinositol polyphosphate phosphohydrolase 3-beta; DIPP3 alpha; Dipp3a; DIPP-3-alpha; DIPP3-alpha; Dipp3b; DIPP-3-beta; hAps2; hDIPP3alpha; MNCb-1696; Nucleoside diphosphate-linked moiety X motif 10; Nucleoside diphosphate-linked moiety X motif 11; nudix (nucleoside diphosphate linked moiety X)-type motif 10; nudix hydrolase 10; nudix motif 10; Nudix motif 11; Nudt10; Nudt11; RGD1564763 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170685 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NFP7 | |
NUDT10 | |
Recombinant protein corresponding to Human NUDT11. Recombinant protein control fragment (Product #RP-104155). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction