Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT12 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306025FR
Description
NUDT12 Polyclonal specifically detects NUDT12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NUDT12 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DKFZp761I172, EC 3.6.1.22, nucleoside diphosphate linked moiety X-type motif 12, Nucleoside diphosphate-linked moiety X motif 12, nudix (nucleoside diphosphate linked moiety X)-type motif 12, Nudix motif 12, peroxisomal NADH pyrophosphatase NUDT12 | |
Produced in rabbits immunized with E. coli-derived Human NUDT12 fragment. | |
0.1 mL | |
Lipid and Metabolism | |
83594 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction