Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP152974
Description
NUDT12 Polyclonal specifically detects NUDT12 in Human samples. It is validated for Western Blot.Specifications
NUDT12 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
DKFZp761I172, EC 3.6.1.22, nucleoside diphosphate linked moiety X-type motif 12, Nucleoside diphosphate-linked moiety X motif 12, nudix (nucleoside diphosphate linked moiety X)-type motif 12, Nudix motif 12, peroxisomal NADH pyrophosphatase NUDT12 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Canine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Chicken: 76%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
Purified |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9BQG2 | |
NUDT12 | |
Synthetic peptides corresponding to NUDT12(nudix (nucleoside diphosphate linked moiety X)-type motif 12) The peptide sequence was selected from the C terminal of NUDT12. Peptide sequence LALAVSTEIKVDKNEIEDARWFTREQVLDVLTKGKQQAFFVPPSRAIAHQ. | |
100 μL | |
Lipid and Metabolism | |
83594 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction