Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDT12 Monoclonal Antibody (3F7)
GREENER_CHOICE

Catalog No. PIMA522698
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522698 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522698 Supplier Invitrogen™ Supplier No. MA522698
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MSSVKRSLKQ EIVTQFHCSA AEGDIAKLTG ILSHSPSLLN ETSENGWTAL MYAARNGHPE IVQFLLEKGC DRSIVNKSRQ TALDIAVFWG YKHIANLLAT AKGGKKPWFL TNEVEECENY FSKTLLDRKS EKRNNSDWLL AKESHPATVF ILFSDLNPLV TLGGNKESFQ QPEVRLCQLN YTDIKDYLAQ PEKITLIFLG VELEIKDKLL NYAGEVPREE EDGLVAWFAL GIDPIAAEEF KQRHENCYFL HPPMPALLQL KEKEAGVVAQ ARSVLAWHSR YKFCPTCGNA TKIEEGGYKR LCLKEDCPSL NGVHNTSYPR VDPVVIMQVI HPDGTKCLLG RQKRFPPGMF TCLAGFIEPG ETIEDAVRRE VEEESGVKVG HVQYVACQPW PMPSSLMIGC LALAVSTEIK VDKNEIEDAR WFTREQVLDV LTKGKQQAFF VPPSRAIAHQ LIKHWIRINP NL.

Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT12
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Nudt12
Gene Accession No. Q9BQG2
Gene Alias 0610016O18Rik; DKFZP761I172; MGC137455 protein; nucleoside diphosphate linked moiety X-type motif 12; nucleoside diphosphate-linked moiety X motif 12; nudix (nucleoside diphosphate linked moiety X)-type motif 12; nudix (nucleoside diphosphnudix (nucleoside diphosphate linked moiety X)-type motif 12ate linked moiety X)-type motif 12; nudix hydrolase 12; nudix motif 12; nudix-type motif 12; NUDT12; Peroxisomal NADH pyrophosphatase NUDT12
Gene Symbols Nudt12
Host Species Mouse
Immunogen NUDT12 (AAH41099, 1 a.a. ~ 462 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83594
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.