Learn More
Invitrogen™ NUDT12 Monoclonal Antibody (OTI3E7)

Description
NUDT12 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | NUDT12 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3E7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Nudt12 |
| Gene Accession No. | Q9BQG2 |
| Gene Alias | 0610016O18Rik; DKFZP761I172; MGC137455 protein; nucleoside diphosphate linked moiety X-type motif 12; nucleoside diphosphate-linked moiety X motif 12; nudix (nucleoside diphosphate linked moiety X)-type motif 12; nudix (nucleoside diphosphnudix (nucleoside diphosphate linked moiety X)-type motif 12ate linked moiety X)-type motif 12; nudix hydrolase 12; nudix motif 12; nudix-type motif 12; NUDT12; Peroxisomal NADH pyrophosphatase NUDT12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.