Learn More
NUDT12 Mouse anti-Human, Clone: OTI2G2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805571

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Specifications
NUDT12 | |
Monoclonal | |
Unconjugated | |
NUDT12 | |
nudix hydrolase 12 | |
Mouse | |
Affinity Chromatography | |
RUO | |
83594 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2G2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BQG2 | |
NUDT12 | |
Human recombit protein fragment corresponding to amino acids 1-251 of human NUDT12 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.