Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDT12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561003
Description
Immunogen sequence: VRREVEEESG VKVGHVQYVA CQPWPMPSSL MIGCLALAVS TEIKVDKNEI EDARWFTREQ VLDVLTKGKQ QAFFVPPSRA IA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.
Specifications
NUDT12 | |
Polyclonal | |
Unconjugated | |
Nudt12 | |
0610016O18Rik; DKFZP761I172; MGC137455 protein; nucleoside diphosphate linked moiety X-type motif 12; nucleoside diphosphate-linked moiety X motif 12; nudix (nucleoside diphosphate linked moiety X)-type motif 12; nudix (nucleoside diphosphnudix (nucleoside diphosphate linked moiety X)-type motif 12ate linked moiety X)-type motif 12; nudix hydrolase 12; nudix motif 12; nudix-type motif 12; NUDT12; Peroxisomal NADH pyrophosphatase NUDT12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83594 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BQG2 | |
Nudt12 | |
Recombinant protein corresponding to Human NUDT12. Recombinant protein control fragment (Product #RP-96866). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction