Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT12 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984333
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4333 Supplier Bioss Supplier No. BS19509R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.

Specifications

Antigen NUDT12
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Nudt12
Gene Accession No. Q9BQG2
Gene Alias 0610016O18Rik; BOS_8526; DKFZP761I172; LOC100354787; nucleoside diphosphate linked moiety X-type motif 12; Nucleoside diphosphate-linked moiety X motif 12; nudix (nucleoside diphosphate linked moiety X)-type motif 12; nudix (nucleoside diphosphnudix (nucleoside diphosphate linked moiety X)-type motif 12ate linked moiety X)-type motif 12; nudix hydrolase 12; Nudix motif 12; nudix-type motif 12; Nudt12; Peroxisomal NADH pyrophosphatase NUDT12; Peroxisomal NADH pyrophosphatase NUDT12-like
Gene Symbols Nudt12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NUDT12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83594
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.