Learn More
Invitrogen™ NUDT13 Monoclonal Antibody (1A9)

Description
Peptide Sequence: SLYCGIACRR KFFWCYRLLS TYVTKTRYLF ELKEDDDACK KAQQTGAFYL FHSLAPLLQT SAHQYLAPRH SLLELERLLG KFGQDAQRIE DSVLIGCSEQ QEAWFALDL.
Specifications
Specifications
| Antigen | NUDT13 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1A9 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | NUDT13 |
| Gene Accession No. | Q86X67 |
| Gene Alias | 4933433B15Rik; LOW QUALITY PROTEIN: nucleoside diphosphate-linked moiety X motif 13; nucleoside diphosphate linked moiety X motif 13; nucleoside diphosphate-linked moiety X motif 13; Nucleoside diphosphate-linked moiety X motif 13-like protein; nudix (nucleoside diphosphate linked moiety X)-type motif 13; nudix hydrolase 13; nudix motif 13; nudix-type motif 13; NUDT13; Protein KiSS-16; Unknown (protein for MGC:140322) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.