Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDT13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569554
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569554 Supplier Invitrogen™ Supplier No. PA569554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 79%; Rabbit: 93%; Rat: 93%.

NUDT13 (Nudix Hydrolase 13) is a Protein Coding gene. Among its related pathways areSynthesis and interconversion of nucleotide di- and triphosphatesandPurine metabolism (REACTOME). Gene Ontology (GO) annotations related to this gene includehydrolase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT13
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NUDT13
Gene Accession No. Q86X67
Gene Alias 4933433B15Rik; LOW QUALITY PROTEIN: nucleoside diphosphate-linked moiety X motif 13; nucleoside diphosphate linked moiety X motif 13; nucleoside diphosphate-linked moiety X motif 13; Nucleoside diphosphate-linked moiety X motif 13-like protein; nudix (nucleoside diphosphate linked moiety X)-type motif 13; nudix hydrolase 13; nudix motif 13; nudix-type motif 13; NUDT13; Protein KiSS-16; Unknown (protein for MGC:140322)
Gene Symbols NUDT13
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human NUDT13.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25961
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.