Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | NUDT14 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NUDT14 Polyclonal specifically detects NUDT14 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
NUDT14 | |
Polyclonal | |
Rabbit | |
metabolism | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
256281 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LTLHYRQNGAQKSWDFMKTHDSVTVLLFNSSRRSLVLVKQFRPAVYAGEVERRFPGSLAAVDQDGPRELQPALPGSAGVTVELCAGLVDQPGLSLEEVAC | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
EC 3.6.1.45, Nucleoside diphosphate-linked moiety X motif 14, nudix (nucleoside diphosphate linked moiety X)-type motif 14, Nudix motif 14, UDPG pyrophosphatase, UDP-sugar diphosphatase, UGPPase, UGPPnucleoside diphosphate linked moiety X-type motif 14, uridine diphosphate glucose pyrophosphatase | |
NUDT14 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title