Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561308
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561308 Supplier Thermo Scientific Supplier No. PA561308
Add to Cart
Add to Cart

NUDT14 Polyclonal Antibody

NUDT14 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT14
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUDT14
Gene Accession No. O95848
Gene Alias 1110030M18Rik; nucleoside diphosphate-linked moiety X motif 14; nudix (nucleoside diphosphate linked moiety X)-type motif 14; nudix hydrolase 14; nudix motif 14; nudix-type motif 14; NUDT14; UDPG pyrophosphatase; UDP-sugar diphosphatase; UGPP; UGPPase; uridine diphosphate glucose pyrophosphatase; Uridine diphosphate glucose pyrophosphatase NUDT14; uridine diphosphate glucose pyrophosphatase NUDT14; uridine diphosphate glucose pyrophosphatase
Gene Symbols NUDT14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUDT14. Recombinant protein control fragment (Product #RP-97612).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256281
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.