Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDT14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570834
Description
This target displays homology in the following species: Cow: 100%; Human: 100%; Mouse: 79%; Pig: 93%; Rat: 86%.
UDP-glucose (UDPG) acts as the sugar donor in numerous glycosylation reactions, including those involved in the production of glycogen. NUDT14 is a UDPG pyrophosphatase (EC 3.6.1.45) that hydrolyzes UDPG to produce glucose 1-phosphate and UMP (Yagi et al., 2003 [PubMed 12429023]).
Specifications
NUDT14 | |
Polyclonal | |
Unconjugated | |
NUDT14 | |
1110030M18Rik; nucleoside diphosphate-linked moiety X motif 14; nudix (nucleoside diphosphate linked moiety X)-type motif 14; nudix hydrolase 14; nudix motif 14; nudix-type motif 14; NUDT14; UDPG pyrophosphatase; UDP-sugar diphosphatase; UGPP; UGPPase; uridine diphosphate glucose pyrophosphatase; Uridine diphosphate glucose pyrophosphatase NUDT14; uridine diphosphate glucose pyrophosphatase NUDT14; uridine diphosphate glucose pyrophosphatase | |
Rabbit | |
Affinity chromatography | |
RUO | |
256281 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O95848 | |
NUDT14 | |
synthetic peptide directed towards the N-terminal region of human NUDT14. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction