Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDT15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558614
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558614 Supplier Invitrogen™ Supplier No. PA558614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKENYHYVTI LMKGEVDVTH DSEPKNVEPE KNESWEWVPW EELPPLDQLF WGLRCLKEQG YDPFKEDLNH LVGYKGNHL Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

Mediates the hydrolysis of some nucleoside diphosphate derivatives. Can degrade 8-oxo-dGTP in vitro, suggesting that it may remove an oxidatively damaged form of guanine (7,8-dihydro-8-oxoguanine) from DNA and the nucleotide pool, thereby preventing misincorporation of 8-oxo-dGTP into DNA thus preventing A:T to C:G transversions. Its substrate specificity in vivo however remains unclear. May have a role in DNA synthesis and cell cycle progression throught the interaction with PCNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT15
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUDT15
Gene Accession No. Q9NV35
Gene Alias 6530403O17; 7,8-dihydro-8-oxoguanine-triphosphatase NUDT15; 8-oxo-dGTPase NUDT15; A730068G11Rik; mMTH2; MTH2; MutT homolog 2; nucleoside diphosphate-linked moiety X motif 15; Nucleoside diphosphate-linked to another moiety X hydrolase 15; Nucleotide triphosphate diphosphatase NUDT15; nudix (nucleoside diphosphate linked moiety X)-type motif 15; nudix hydrolase 15; Nudix motif 15; nudix-type motif 15; NUDT15; NUDT15D; probable 7,8-dihydro-8-oxoguanine triphosphatase NUDT15; probable 8-oxo-dGTP diphosphatase NUDT15; RP11-90M2.1
Gene Symbols NUDT15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUDT15. Recombinant protein control fragment (Product #RP-102861).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55270
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.