Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDT15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589643
Description
Immunogen sequence: MTASAQPRGR RPGVGVGVVV TSCKHPRCVL LGKRKGSVGA GSFQLPGGHL EFGETWEECA QRETWEEAAL HLKNVHFASV VNSFIEKENY HYVTILMKGE VDVTHDSEPK NVEPEKNESW EWVPWEELPP LDQLFWGLRC LKEQGYDPFK EDLNHLVGYK GNHL; Positive Samples: Mouse spleen.
Mediates the hydrolysis of some nucleoside diphosphate derivatives. Can degrade 8-oxo-dGTP in vitro, suggesting that it may remove an oxidatively damaged form of guanine (7,8-dihydro-8-oxoguanine) from DNA and the nucleotide pool, thereby preventing misincorporation of 8-oxo-dGTP into DNA thus preventing A:T to C:G transversions. Its substrate specificity in vivo however remains unclear. May have a role in DNA synthesis and cell cycle progression throught the interaction with PCNA.
Specifications
NUDT15 | |
Polyclonal | |
Unconjugated | |
NUDT15 | |
6530403O17; 7,8-dihydro-8-oxoguanine-triphosphatase NUDT15; 8-oxo-dGTPase NUDT15; A730068G11Rik; mMTH2; MTH2; MutT homolog 2; nucleoside diphosphate-linked moiety X motif 15; Nucleoside diphosphate-linked to another moiety X hydrolase 15; Nucleotide triphosphate diphosphatase NUDT15; nudix (nucleoside diphosphate linked moiety X)-type motif 15; nudix hydrolase 15; Nudix motif 15; nudix-type motif 15; NUDT15; NUDT15D; probable 7,8-dihydro-8-oxoguanine triphosphatase NUDT15; probable 8-oxo-dGTP diphosphatase NUDT15; RP11-90M2.1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
214254, 290365 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BG93 | |
NUDT15 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-164 of human NUDT15 (NP_0607531). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction