Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT16 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2854140.1ML
Description
NUDT16 Polyclonal specifically detects NUDT16 in Mouse samples. It is validated for Western Blot.Specifications
NUDT16 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
131870 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
EC 3.6.1, EC 3.6.1.-, FLJ31265, FLJ34034, FLJ36248, Nucleoside diphosphate-linked moiety X motif 16, nudix (nucleoside diphosphate linked moiety X)-type motif 16, Nudix motif 16, U8 snoRNA-binding protein H29K, U8 snoRNA-decapping enzyme | |
The immunogen is a synthetic peptide directed towards the N-terminal region of NUDT16 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction