Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDT17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569539
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569539 Supplier Invitrogen™ Supplier No. PA569539
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rat: 86%.

Probably mediates the hydrolysis of some nucleoside diphosphate derivatives.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT17
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NUDT17
Gene Accession No. P0C025
Gene Alias 2410015C20Rik; nucleoside diphosphate-linked moiety X motif 17; nudix (nucleoside diphosphate linked moiety X)-type motif 17; nudix hydrolase 17; nudix motif 17; NUDT17; RGD1565469
Gene Symbols NUDT17
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human NUDT17.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200035
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.