Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT18 Antibody (OTI5D7), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273110PECY55
Description
NUDT18 Monoclonal specifically detects NUDT18 in Human samples. It is validated for Flow Cytometry.Specifications
NUDT18 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
NUDT18 | |
Full length recombinant protein of human NUDT18 (NP_079091) produced in HEK293T cells. | |
0.1 mL | |
79873.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI5D7 | |
Flow Cytometry | |
EC 3.6.1, EC 3.6.1.-, FLJ22494, nucleoside diphosphate-linked moiety X motif 18, nudix (nucleoside diphosphate linked moiety X)-type motif 18, Nudix motif 18 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction