Learn More
NUDT18 Mouse anti-Human, Clone: OTI5F9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503865

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the Nudix hydrolase family. Nudix hydrolases eliminate potentially toxic nucleotide metabolites from the cell and regulate the concentrations and availability of many different nucleotide substrates, cofactors, and signaling molecules. This protein contains a Nudix hydrolase domain and hydrolyzes oxidized forms of guanosine and deoxyguanosine diphosphates.Specifications
NUDT18 | |
Monoclonal | |
Unconjugated | |
NUDT18 | |
MTH3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
79873 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI5F9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q6ZVK8 | |
NUDT18 | |
Full length human recombit protein of human NUDT18 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.