Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT19 Antibody (SR1715), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | NUDT19 |
---|---|
Clone | SR1715 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
NUDT19 Monoclonal antibody specifically detects NUDT19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
NUDT19 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
390916 | |
IgG | |
Affinity purified |
SR1715 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
EC 3.6.1, Nucleoside Diphosphate-Linked Moiety X Motif 19, Mitochondrial, Nudix (Nucleoside Diphosphate Linked Moiety X)-Type Motif 19, Nudix Motif 19, RP2 | |
A synthesized peptide derived from human NUDT19 (Uniprot #: A8MXV4) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title