Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDT19 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551352

Catalog No. PA551352


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total NUDT19 protein.

NUDT19 is a coenzyme A diphosphatase that mediates the hydrolysis of a wide range of CoA esters, including choloyl-CoA and branched-chain fatty-acyl-CoA esters. At low substrate concentrations medium and long-chain fatty-acyl-CoA esters, NUDT19 are the primary substrates.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NUDT19
Polyclonal
Unconjugated
NUDT19
CTC-379B2.4; nucleoside diphosphate-linked moiety X motif 19; nucleoside diphosphate-linked moiety X motif 19, mitochondrial; nudix (nucleoside diphosphate linked moiety X)-type motif 19; nudix hydrolase 19; nudix motif 19; NUDT19; RP2
Rabbit
Antigen Affinity Chromatography
RUO
390916
-20°C
Liquid
Western Blot
1.5 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
A8MXV4
NUDT19
Synthetic peptide of human NUDT19.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.