Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP257066 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP257066 100 μL
NB394153 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP257066 Supplier Novus Biologicals Supplier No. NBP257066
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUDT2 Polyclonal specifically detects NUDT2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NUDT2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias Ap4A hydrolase, Ap4A hydrolase 1, Ap4Aase, APAH1Diadenosine 5'-5'''-P1, bis(5'-nucleosyl)-tetraphosphatase (asymmetrical), bis(5'-nucleosyl)-tetraphosphatase [asymmetrical], diadenosine 5'-5''-P1, Diadenosine tetraphosphatase, EC 3.6.1.17, MGC10404, Nucleoside diphosphate-linked moiety X motif 2, nudix (nucleoside diphosphate linked moiety X)-type motif 2, Nudix motif 2, P4-tetraphosphate asymmetrical hydrolase, P4-tetraphosphate pyrophosphohydrolase
Gene Symbols NUDT2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MALRACGLIIFRRCLIPKVDNNAIEFLLLQASDGIHHWTPPKGHVEPGEDDLETALR
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 318
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.