Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT21 Antibody (OTI13H1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273113R
Description
NUDT21 Monoclonal specifically detects NUDT21 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
NUDT21 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NUDT21 | |
Recombinant protein fragment corresponding to amino acids 2-227 of human NUDT21 (NP_008937) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI13H1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CFIm25, CFIM25CPSF 25 kDa subunit, cleavage and polyadenylation specific factor 5, 25 kD subunit, cleavage and polyadenylation specific factor 5, 25 kDa, Cleavage and polyadenylation specificity factor 25 kDa subunit, cleavage and polyadenylation specificity factor subunit 5, CPSF25, CPSF5DKFZp686H1588, Nucleoside diphosphate-linked moiety X motif 21, nudix (nucleoside diphosphate linked moiety X)-type motif 21, Nudix motif 21, pre-mRNA cleavage factor Im (25kD), Pre-mRNA cleavage factor Im 25 kDa subunit, pre-mRNA cleavage factor Im, 25kD subunit | |
Mouse | |
Affinity Purified | |
Proteases & Other Enzymes | |
11051.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction