Learn More
Invitrogen™ NUDT21 Polyclonal Antibody

Description
Antibody detects endogenous levels of total NUDT21.
Specifications
Specifications
| Antigen | NUDT21 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | NUDT21 |
| Gene Accession No. | O43809, Q4KM65, Q9CQF3 |
| Gene Alias | 25kDa; 3110048P04Rik; 5730530J16Rik; AU014860; AW549947; CFIM25; cleavage and polyadenylation specific factor 5; cleavage and polyadenylation specific factor 5, 25 kD subunit; cleavage and polyadenylation specific factor 5, 25 kDa; cleavage and polyadenylation specificity factor 25 kDa subunit; Cleavage and polyadenylation specificity factor subunit 5; cleavage factor Im complex 25 kDa subunit; CPSF 25 kDa subunit; CPSF25; cpsf5; nucleoside diphosphate-linked moiety X motif 21; nudix (nucleoside diphosphate linked moiety X)-type motif 21; nudix hydrolase 21; Nudix motif 21; Nudt21; pre-mRNA cleavage factor Im (25kD); pre-mRNA cleavage factor Im 25 kDa subunit; Pre-mRNA cleavage factor Im 68 kDa subunit; pre-mRNA cleavage factor Im, 25kD subunit; similar to cleavage and polyadenylation specific factor 5, 25 kDa; zgc:63966 |
| Gene Symbols | NUDT21 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.