Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    NUDT22 Antibody, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25538125UL
Description
NUDT22 Polyclonal specifically detects NUDT22 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| NUDT22 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL | |
| MGC13045, nucleoside diphosphate-linked moiety X motif 22, nudix (nucleoside diphosphate linked moiety X)-type motif 22, Nudix motif 22 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 84304 | |
| Human | |
| IgG | 
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| NUDT22 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DPEVTLLLQCPGGGLPQEQIQAELSPAHDRRPLPGGDEAITAIWETRLKAQPWLFDAPKFRLHSATLAPIGSRGPQLLLRLGLT | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            