Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDT22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584156
Description
Immunogen sequence: DPEVTLLLQC PGGGLPQEQI QAELSPAHDR RPLPGGDEAI TAIWETRLKA QPWLFDAPKF RLHSATLAPI GSRGPQLLLR LGLT.
NUDT22 is a protein coding gene. Hydrolyzes UDP-glucose to glucose 1-phosphate and UMP and UDP-galactose to galactose 1-phosphate and UMP. Preferred substrate is UDP-glucose.
Specifications
NUDT22 | |
Polyclonal | |
Unconjugated | |
NUDT22 | |
MGC13045; Nucleoside diphosphate-linked moiety X motif 22; NUD22; nudix (nucleoside diphosphate linked moiety X)-type motif 22; nudix hydrolase 22; nudix motif 22; NUDT22; PP11246; UDPG pyrophosphatase; Uridine diphosphate glucose pyrophosphatase NUDT22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84304 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRQ3 | |
NUDT22 | |
Recombinant protein corresponding to Human NUDT22. Recombinant protein control fragment (Product #RP-106465). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction