Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | NUDT3 |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NUDT3 Polyclonal antibody specifically detects NUDT3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
NUDT3 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2), 40% Glycerol | |
11165 | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Diadenosine 5'-5'''-P1, diphosphoinositol polyphosphate phosphohydrolase 1, DIPP-1, DIPPDIPP1, EC 3.6.1.-, EC 3.6.1.52, Nucleoside diphosphate-linked moiety X motif 3, nudix (nucleoside diphosphate linked moiety X)-type motif 3, Nudix motif 3, P6-hexaphosphate hydrolase 1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LRQGYSANNGTPVVATTYSVSAQSSMSGIR | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title