Learn More
Invitrogen™ NUDT3 Monoclonal Antibody (3C5)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA521285
Catalog No. PIMA521285
Only null left
Add to Cart
Description
Description
Peptide Sequence: MMKLKSNQTR TYDGDGYKKR AACLCFRSES EEEVLLVSSS RHPDRWIVPG GGMEPEEEPS VAAVREVCEE AGVKGTLGRL VGIFENQERK HRTYVYVLIV TEVLEDWEDS VNIGRKREWF KIEDAIKVLQ YHKPVQASYF ETLRQGYSAN NGTPVVATTY SVSAQSSMSG IR.
NUDT3 belongs to the MutT, or Nudix, protein family. Nudix proteins act as homeostatic checkpoints at important stages in nucleoside phosphate metabolic pathways, guarding against elevated levels of potentially dangerous intermediates, like 8-oxo-dGTP, which promotes AT-to-CG transversions (Safrany et al., 1998 ).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NUDT3
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
O95989
NUDT3
NUDT3 (AAH07727, 1 a.a. ~ 172 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG1 κ
ELISA, Western Blot
3C5
Unconjugated
NUDT3
1110011B09Rik; AA960325; diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 1; Diphosphoinositol polyphosphate phosphohydrolase 1; Dipp; DIPP1; DIPP-1; Nucleoside diphosphate-linked moiety X motif 3; nudix (nucleoside diphosphate linked moiety X)-type motif 3; nudix (nucleotide diphosphate linked moiety X)-type motif 3; nudix hydrolase 3; nudix motif 3; NUDT3
Mouse
Affinity chromatography
RUO
11165
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.