Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530435
Description
Recommended positive controls: IMR32. Predicted reactivity: Mouse (97%), Rat (96%), Dog (99%), Pig (99%), Chicken (95%), Rhesus Monkey (100%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NUDT3 belongs to the MutT, or Nudix, protein family. Nudix proteins act as homeostatic checkpoints at important stages in nucleoside phosphate metabolic pathways, guarding against elevated levels of potentially dangerous intermediates, like 8-oxo-dGTP, which promotes AT-to-CG transversions (Safrany et al., 1998 ).
Specifications
NUDT3 | |
Polyclonal | |
Unconjugated | |
NUDT3 | |
1110011B09Rik; AA960325; diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 1; Diphosphoinositol polyphosphate phosphohydrolase 1; Dipp; DIPP1; DIPP-1; Nucleoside diphosphate-linked moiety X motif 3; nudix (nucleoside diphosphate linked moiety X)-type motif 3; nudix (nucleotide diphosphate linked moiety X)-type motif 3; nudix hydrolase 3; nudix motif 3; NUDT3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11165 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O95989 | |
NUDT3 | |
Recombinant protein encompassing a sequence within the center region of human NUDT3. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction