Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDT4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551213

Catalog No. PA551213


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total NUDT4 protein.

Cleaves a beta-phosphate from the diphosphate groups in PP-InsP5 (diphosphoinositol pentakisphosphate), PP-InsP4 and [PP]2-InsP4 (bisdiphosphoinositol tetrakisphosphate), suggesting that it may play a role in signal transduction. Also able to catalyze the hydrolysis of dinucleoside oligophosphate Ap6A, but not Ap5A. The major reaction products are ADP and p4a from Ap6A. Also able to hydrolyze 5-phosphoribose 1-diphosphate. Does not play a role in U8 snoRNA decapping activity. Binds U8 snoRNA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NUDT4
Polyclonal
Unconjugated
NUDT4
4933436C10Rik; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 2; diphosphoinositol polyphosphate phosphohydolase type II; diphosphoinositol polyphosphate phosphohydrolase 2; Diphosphoinositol polyphosphate phosphohydrolase NUDT4B; diphosphoinositol polyphosphate phosphohydrolase type 2; DIPP2; DIPP-2; DIPP2alpha; DIPP-2B; DIPP2beta; DRCF-5; HDCMB47P; KIAA0487; Nucleoside diphosphate-linked moiety X motif 4; Nucleoside diphosphate-linked moiety X motif 4B; nudix (nucleoside diphosphate linked moiety X)-type motif 4; nudix hydrolase 4; Nudix hydrolase 4B; Nudix motif 4; Nudix motif 4B; nudix-type motif 4; NUDT4; NUDT4B; rDIPP2
Rabbit
Antigen affinity chromatography
RUO
11163
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9NZJ9
NUDT4
Full length fusion protein of human NUDT4.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.