Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587942 Supplier Thermo Scientific Supplier No. PA587942
Add to Cart
Add to Cart

NUDT5 Polyclonal Antibody

NUDT5 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NUDT5
Gene Accession No. Q6AY63, Q9JKX6, Q9UKK9
Gene Alias 8-oxo-dGDP phosphatase; ADP-sugar pyrophosphatase; HSPC115; Nuclear ATP-synthesis protein NUDIX5; Nucleoside diphosphate-linked moiety X motif 5; nudix (nucleoside diphosphate linked moiety X)-type motif 5; nudix hydrolase 5; nudix motif 5; NUDIX5; nudix-type motif 5; NUDT5; YSA1; YSA1H; YSAH1
Gene Symbols NUDT5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-219 of human NUDT5 (NP_0548612).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11164, 361274, 53893
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.