Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDT5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554091
Description
Immunogen sequence: SPAVCMDPGL SNCTIHIVTV TINGDDAENA RPKPKPGDGE FVEVISLPKN DLLQRLDALV AEEHLTVDAR VYSYALALKH ANAKPFEVPF LKF Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.
Enzyme that can either act as an ADP-sugar pyrophosphatase in absence of diphosphate or catalyze the synthesis of ATP in presence of diphosphate. In absence of diphosphate, hydrolyzes with similar activities various modified nucleoside diphosphates such as ADP-ribose, ADP-mannose, ADP-glucose, 8-oxo-GDP and 8-oxo-dGDP. Can also hydrolyze other nucleotide sugars with low activity. In presence of diphosphate, mediates the synthesis of ATP in the nucleus by catalyzing the conversion of ADP-ribose to ATP and ribose 5-phosphate. Nuclear ATP synthesis takes place when dephosphorylated at Thr-45. Nuclear ATP generation is required for extensive chromatin remodeling events that are energy-consuming. Does not play a role in U8 snoRNA decapping activity. Binds U8 snoRNA.
Specifications
NUDT5 | |
Polyclonal | |
Unconjugated | |
NUDT5 | |
8-oxo-dGDP phosphatase; ADP-sugar pyrophosphatase; HSPC115; Nuclear ATP-synthesis protein NUDIX5; Nucleoside diphosphate-linked moiety X motif 5; nudix (nucleoside diphosphate linked moiety X)-type motif 5; nudix hydrolase 5; nudix motif 5; NUDIX5; nudix-type motif 5; NUDT5; YSA1; YSA1H; YSAH1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11164, 361274 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6AY63, Q9UKK9 | |
NUDT5 | |
Recombinant protein corresponding to Human NUDT5. Recombinant protein control fragment (Product #RP-92485). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction