Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDT5 Recombinant Rabbit Monoclonal Antibody (10D2)
GREENER_CHOICE

Catalog No. FEMA550039
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550039 Supplier Invitrogen™ Supplier No. MA550039
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

NUDT5 Recombinant Monoclonal Antibody for ELISA, Flow, Western Blot

Enzyme that can either act as an ADP-sugar pyrophosphatase in absence of diphosphate or catalyze the synthesis of ATP in presence of diphosphate. In absence of diphosphate, hydrolyzes with similar activities various modified nucleoside diphosphates such as ADP-ribose, ADP-mannose, ADP-glucose, 8-oxo-GDP and 8-oxo-dGDP. Can also hydrolyze other nucleotide sugars with low activity. In presence of diphosphate, mediates the synthesis of ATP in the nucleus by catalyzing the conversion of ADP-ribose to ATP and ribose 5-phosphate. Nuclear ATP synthesis takes place when dephosphorylated at Thr-45. Nuclear ATP generation is required for extensive chromatin remodeling events that are energy-consuming. Does not play a role in U8 snoRNA decapping activity. Binds U8 snoRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT5
Applications ELISA, Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone 10D2
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NUDT5
Gene Accession No. Q9UKK9
Gene Alias 8-oxo-dGDP phosphatase; ADP-sugar pyrophosphatase; HSPC115; Nuclear ATP-synthesis protein NUDIX5; Nucleoside diphosphate-linked moiety X motif 5; nudix (nucleoside diphosphate linked moiety X)-type motif 5; nudix hydrolase 5; nudix motif 5; NUDIX5; nudix-type motif 5; NUDT5; YSA1; YSA1H; YSAH1
Gene Symbols NUDT5
Host Species Rabbit
Immunogen A synthesized peptide derived from human NUDT5.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11164
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.