Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT6 Monoclonal Antibody (OTI9D5), TrueMAB™, OriGene
SDP

Catalog No. 502541626
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1626 30 μL
50-167-3393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1626 Supplier OriGene Supplier No. TA502331S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NUDT6 Monoclonal antibody specifically detects NUDT6 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen NUDT6
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI9D5
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NUDT6
Gene Accession No. P53370
Gene Alias antisense basic fibroblast growth factor; Antisense basic fibroblast growth factor B; anti-sense basic fibroblast growth factor B; ASFGF2; Asfgf2b; FGF2AS; FGF-AS; Gfg; GFG1; GFG-1; Nucleoside diphosphate-linked moiety X motif 6; nudix (nucleoside diphosphate linked moiety X)-type motif 6; nudix hydrolase 6; nudix motif 6; Nudt6; Protein GFG
Host Species Mouse
Immunogen Full length human recombinant protein of human NUDT6 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11162
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.