Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDT7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559837 Supplier Invitrogen™ Supplier No. PA559837
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSRLGLPEEP VRNSLLDDAK ARLRKYDIGG KYSHLPYNKY SVLLPLVAKE GKLHLLFTVR SEKLRRAPGE VCFPGGKRDP TD Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 68%.

The protein encoded by this gene is a member of the Nudix hydrolase family. Nudix hydrolases eliminate potentially toxic nucleotide metabolites from the cell and regulate the concentrations and availability of many different nucleotide substrates, cofactors, and signaling molecules. Alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUDT7
Gene Accession No. P0C024
Gene Alias 1300007B24Rik; 2210404C19Rik; coenzyme A diphosphatase; nucleoside diphosphate-linked moiety X motif 7; nudix (nucleoside diphosphate linked moiety X)-type motif 7; nudix hydrolase 7; nudix motif 7; Nudt7; Peroxisomal coenzyme A diphosphatase NUDT7; peroxisomal coenzyme A diphosphatase NUDT7-like
Gene Symbols NUDT7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUDT7. Recombinant protein control fragment (Product #RP-98721).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283927
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.