Learn More
Invitrogen™ NUDT8 Polyclonal Antibody

Description
Immunogen sequence: KCDPADQDVV HTALRETREE LGLAVPEEHV WGLLRPVYDP QKATVVPVLA GVGPLDPQSL RPNSEEVD Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 79%.
Specifications
Specifications
| Antigen | NUDT8 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Nudt8 |
| Gene Accession No. | Q8WV74 |
| Gene Alias | 2310039H17Rik; Nucleoside diphosphate-linked moiety X motif 8; nucleoside diphosphate-linked moiety X motif 8, mitochondrial; nudix (nucleoside diphosphate linked moiety X)-type motif 8; nudix hydrolase 8; nudix motif 8; nudix-type motif 8; Nudt8 |
| Gene Symbols | Nudt8 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.