Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDT8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559493
Description
Immunogen sequence: EVFALPLAHL LQTQNQGYTH FCRGGHFRYT LPVFLHGPHR VWGLTAVITE FALQLLAPGT YQPRLAGLTC SGAEGLARPK QPLASPCQAS STPGLNKGL Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 56%.
Probably mediates the hydrolysis of some nucleoside diphosphate derivatives.
Specifications
NUDT8 | |
Polyclonal | |
Unconjugated | |
Nudt8 | |
2310039H17Rik; Nucleoside diphosphate-linked moiety X motif 8; nucleoside diphosphate-linked moiety X motif 8, mitochondrial; nudix (nucleoside diphosphate linked moiety X)-type motif 8; nudix hydrolase 8; nudix motif 8; nudix-type motif 8; Nudt8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
254552 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WV74 | |
Nudt8 | |
Recombinant protein corresponding to Human NUDT8. Recombinant protein control fragment (Product #RP-97453). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction