Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT9 Antibody (OTI7A12), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273126H
Description
NUDT9 Monoclonal specifically detects NUDT9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NUDT9 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
NUDT9 | |
Full length recombinant protein of human NUDT9 (NP_076952) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI7A12 | |
Western Blot, Immunohistochemistry | |
ADP-ribose diphosphatase, ADP-ribose phosphohydrolase, ADP-ribose pyrosphosphatase NUDT9, ADPR-PPase, EC 3.6.1.13, MGC3037, mitochondrial, nucleoside diphosphate linked moiety X-type motif 9, nudix (nucleoside diphosphate linked moiety X)-type motif 9, Nudix motif 9, NUDT10 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
53343.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction