Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT9 Monoclonal Antibody (OTI 7F12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503235

Description
TRAC-1 is a 232 amino acid protein containing an N-terminal RING-type zinc finger domain. It functions as a positive regulator in the T-cell receptor signaling pathway, thus leading to T-cell activation. It mediates ubiquitination and subsequent proteasomal degradation of target proteins.TRAC-1 is expressed in lymphoid tissues, including bone marrow, spleen, thymus as well as in other tissues. It is predomitly expressed in the CD4+ and CD8+ T-cells, suggesting that it is preferentially confined to T-cells.Specifications
NUDT9 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9BW91 | |
NUDT9 | |
Full length human recombit protein of human NUDT9 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
OTI 7F12 | |
Unconjugated | |
NUDT9 | |
NUDT10 | |
Mouse | |
Affinity Chromatography | |
RUO | |
53343 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction