Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDT9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541085
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541085 Supplier Invitrogen™ Supplier No. PA541085
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LEAGDDAGKV KWVDINDKLK LYASHSQFIK LVAEKRDAHW SEDSEADCHA Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 86%; Zebrafish: 77%.

YBX1 binds to splice sites in pre-mRNA and regulates splice site selection. This protein binds and stabilizes cytoplasmic mRNA and contributes to the regulation of translation by modulating the interaction between the mRNA and eukaryotic initiation factors. It binds to promoters that contain a Y-box (5'-CTGATTGGCCAA-3'), such as HLA class II genes. It regulates the transcription of numerous genes and promotes separation of DNA strands that contain mismatches or are modified by cisplatin. It has endonucleolytic activity and can introduce nicks or breaks into double-stranded DNA (in vitro), and it may play a role in DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT9
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Nudt9
Gene Accession No. Q9BW91
Gene Alias 1190002C07Rik; Adenosine diphosphoribose pyrophosphatase; ADP-ribose diphosphatase; ADP-ribose phosphohydrolase; ADP-ribose pyrophosphatase; ADP-ribose pyrophosphatase, mitochondrial; ADP-ribose pyrosphosphatase NUDT9; ADPR-PPase; AI462474; fu33g07; hypothetical protein LOC406661; mitochondrial ADP-ribose pyrophosphatase-like protein; nucleoside diphosphate linked moiety X-type motif 9; nucleoside diphosphate-linked moiety X motif 9; nudix (nucleoside diphosphate linked moiety X)-type motif 9; nudix hydrolase 9; nudix motif 9; nudix -type motif 9; nudix-type motif 9; NUDT10; Nudt9; PSEC0099; sb:cb392; UNQ3012/PRO9771; wu:fj16b09; wu:fu33g07; zgc:63924
Gene Symbols Nudt9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human NUDT9 (aa 300-349).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53343
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.