Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDT9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557869
Description
Immunogen sequence: AWMETEAVNY HDETGEIMDN LMLEAGDDAG KVKWVDINDK LKLYASHSQF IKLVAEKRDA HWSEDSEADC HAL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
YBX1 binds to splice sites in pre-mRNA and regulates splice site selection. This protein binds and stabilizes cytoplasmic mRNA and contributes to the regulation of translation by modulating the interaction between the mRNA and eukaryotic initiation factors. It binds to promoters that contain a Y-box (5'-CTGATTGGCCAA-3'), such as HLA class II genes. It regulates the transcription of numerous genes and promotes separation of DNA strands that contain mismatches or are modified by cisplatin. It has endonucleolytic activity and can introduce nicks or breaks into double-stranded DNA (in vitro), and it may play a role in DNA repair.
Specifications
NUDT9 | |
Polyclonal | |
Unconjugated | |
Nudt9 | |
1190002C07Rik; Adenosine diphosphoribose pyrophosphatase; ADP-ribose diphosphatase; ADP-ribose phosphohydrolase; ADP-ribose pyrophosphatase; ADP-ribose pyrophosphatase, mitochondrial; ADP-ribose pyrosphosphatase NUDT9; ADPR-PPase; AI462474; fu33g07; hypothetical protein LOC406661; mitochondrial ADP-ribose pyrophosphatase-like protein; nucleoside diphosphate linked moiety X-type motif 9; nucleoside diphosphate-linked moiety X motif 9; nudix (nucleoside diphosphate linked moiety X)-type motif 9; nudix hydrolase 9; nudix motif 9; nudix -type motif 9; nudix-type motif 9; NUDT10; Nudt9; PSEC0099; sb:cb392; UNQ3012/PRO9771; wu:fj16b09; wu:fu33g07; zgc:63924 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
53343 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BW91 | |
Nudt9 | |
Recombinant protein corresponding to Human NUDT9. Recombinant protein control fragment (Product #RP-96735). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction