Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nuf2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$330.00 - $547.00
Specifications
Antigen | Nuf2 |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Nuf2 Polyclonal antibody specifically detects Nuf2 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Nuf2 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS, pH 7.2, 40% glycerol | |
83540 | |
IgG | |
Affinity purified |
Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
cancer/testis antigen 106, CDCA1hsNuf2, cell division cycle associated 1, Cell division cycle-associated protein 1, CT106, hNuf2, kinetochore protein Nuf2, NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae), NUF2RhNuf2R | |
This antibody was developed against Recombinant Protein corresponding to amino acids: YGDSVDCLPSCQLEVQLYQKKIQDLSDNREKLASILKESLNLEDQIESDESELKKLKTEENSFKRLMIVKKE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title